Masterflex® Reducer Fittings, Compression, Straight, Avantor®
Proveedor: Avantor Fluid Handling
Masterflex®
MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP
MFLX06381-72EA 943 MXN
MFLX06381-72 MFLX06381-70 MFLX30803-09 MFLX30805-08 MFLX30805-09 MFLX30805-06 MFLX30805-07 MFLX06381-76 MFLX06381-74 MFLX30805-10 MFLX06382-19 MFLX06382-18 MFLX06382-17 MFLX06381-05 MFLX30815-00
Masterflex® Reducer Fittings, Compression, Straight, Avantor®
Racores y conectores
Excellent choice to connect rigid tubing systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rates to 220 psi at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).