Masterflex® L/S® High-Performance Precision Pump Tubing, FDA-Compliant Viton®, Avantor®
Proveedor: Avantor Fluid Handling
Masterflex®
96412-24 96412-36 96412-35 96412-15
MFLX96412-24EA 13093 MXN
MFLX96412-24 MFLX96412-35 MFLX96412-36 MFLX96412-15
Masterflex® L/S® High-Performance Precision Pump Tubing, FDA-Compliant Viton®, Avantor®
Tuberías Tubería de bomba peristáltica
Good for food and lab applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work
Characteristics: Good chemical resistance, opaque black
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.