Masterflex® Transfer Tubing, Versilon™ 2001, Avantor®
Proveedor: Avantor Fluid Handling
Masterflex®
AD300007-CP AD300017-CP AD300027-CP AD300038-CP AD300012-CP
MFLX95716-06EA 6854 MXN
MFLX95716-06 MFLX95716-29 MFLX95716-14 MFLX95716-11 MFLX95716-21
Masterflex® Transfer Tubing, Versilon™ 2001, Avantor®
Tuberías Tubería de bomba peristáltica
Ultra-smooth inert inner bore provides excellent chemical resistance.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165 °F (-46 to 73 °C). Can be sterilized with ethylene oxide or autoclave.