Masterflex® Metering Pump Systems, Avantor®
Proveedor: Avantor Fluid Handling
Masterflex®
T15N302X000V03 T15N102X000V02 T15N402X000V03 T15N002X000V02
MFLX76303-00EA 21017 MXN
MFLX76303-00 MFLX76303-10 MFLX76303-20 MFLX76303-30
Masterflex® Metering Pump Systems, Avantor®
Bombas Bombas dosificadoras de membrana
Complete, self-contained system for metering precise amounts of fluid.
  • Maximum flow up to 480 ml/min│Durable polyethylene tank with extra heavy wall thickness for enhanced strength│Available in 15 and 30 gallon capacities
  • Unit fits easily in any out-of-the-way corner
  • Top-mounted, multi-position tank opening — leave open, closed or vented
Metering, injector features heavy-duty, double ball valve pump head — enhanced priming capabilities make it exceptional for purging air and gas.