Masterflex® Fittings, Compression Nut Assembly with Ferrule, Avantor®
Proveedor: Avantor Fluid Handling
Masterflex®
MAKE-UP MAKE-UP MAKE-UP MAKE-UP
MFLX06380-11EA 603 MXN
MFLX06380-11 MFLX06380-21 MFLX06380-31 MFLX06380-41
Masterflex® Fittings, Compression Nut Assembly with Ferrule, Avantor®
Racores y conectores
Can handle pressures up to 225 psi.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 220 psi maximum at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).