Masterflex® Computer Software and Cables, Avantor®
Proveedor: Avantor Fluid Handling
Masterflex®
7550-64
MFLX07550-64EA 4030 MXN
MFLX07550-64
Masterflex® Computer Software and Cables, Avantor®
Homogenizadores Mezcladores estándar de laboratorio
Easily connect your Masterflex® L/S® computer-compatible drive to a PC.
  • Featuring RS-232 connections with 8 foot cable length
Monitor and control your Servodyne mixer system from an IBM® or 100% compatible computer via a computer interface. Software programs let you to control up to 16 Servodyne mixers and Masterflex® computer-compatible pumps. Various application programs let you dedicate your automated setup for mixing and diluting, sequence control, flow proportioning, reversible batch dispensing, delivering single and multiple slope gradients, or customizing procedures for specialized applications.