Hayward Bag Filters, Glass-Reinforced Polypropylene
Proveedor: Hayward
PO005G2PRSH PO100G2PRSH PO025G1PRSH
MFLX29990-74EA 11476 MXN
MFLX29990-74 MFLX29990-82 MFLX29990-58
Hayward Bag Filters, Glass-Reinforced Polypropylene
Filtros Bag Filter Systems
Conveniently collect all filtered particles within the bag.
  • Bags are constructed of glass-reinforced polypropylene (PP)
Hayward(R) bags are made from nonwoven polypropylene felt. The single-length bags have a surface area of 2.0 ft2; double length bags have a surface area of 4.1 ft2. Bags are available in mulitple pore sizes.