Centrifugal Multi-Purpose Separator/Extractor Weirs
Supplier: CINC INDUSTRIES INC
Certificates
About this item
Mix, react, and/or separate liquid-liquid mixtures in a single device.
- Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
- Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
Weirs are available in various diameters. Use the large diameter weirs in applications with high-viscosity liquids.
- Environmentally Preferable:
Specifications
- Warranty:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
Specifications
Product Information
- Catalog #Supplier #ØAvailabilityPrice
- Centrifugal Multi-Purpose Separator/Extractor Weir; 1.125" DiameterCentrifugal Multi-Purpose Separator/Extractor Weir; 1.125" Diameter02W1.125B-22.8575 cmLog in for availabilityEach (1UN)$754.00Specifications:
More
Specifications
Environmentally PreferableWarranty2 years - Centrifugal Multi-Purpose Separator/Extractor Weir; 1.100" DiameterCentrifugal Multi-Purpose Separator/Extractor Weir; 1.100" Diameter02W1.100B-22.794 cmLog in for availabilityEach (1UN)$754.00Specifications:
More
Specifications
Environmentally PreferableWarranty2 years - Centrifugal Multi-Purpose Separator/Extractor Weir; 1.175" DiameterCentrifugal Multi-Purpose Separator/Extractor Weir; 1.175" Diameter02W1.175B-22.9845 cmLog in for availabilityEach (1UN)$754.00Specifications:
More
Specifications
Environmentally PreferableWarranty2 years - Centrifugal Multi-Purpose Separator/Extractor Weir; 1.150" DiameterCentrifugal Multi-Purpose Separator/Extractor Weir; 1.150" Diameter02W1.150B-22.921 cmLog in for availabilityEach (1UN)$754.00Specifications:
More
Specifications
Environmentally PreferableWarranty2 years - Centrifugal Multi-Purpose Separator/Extractor Weir; 1.075" DiameterCentrifugal Multi-Purpose Separator/Extractor Weir; 1.075" Diameter02W1.075B-22.7305 cmLog in for availabilityEach (1UN)$754.00Specifications:
More
Specifications
Environmentally PreferableWarranty2 years