Iron Reduction Cartridge for "Big Blue" housing
Proveedor: Pentair
155263-03
MFLX29830-62EA 4463 MXN
MFLX29830-62
Iron Reduction Cartridge for "Big Blue" housing
Filtros Cartuchos de filtros
Reduce iron and metallic taste in water.
  • Designed with Permanganese filter media for iron reduction
Iron Reduction Cartridge provides an easy yet effective way to reduce iron in water. Reduces the metallic taste of water and helps eliminate stains on plumbing fixtures. Ideal for drinking water.