Saint Gobain® Plastic Luer-to-Sanitary Adapters
Proveedor: Saint-Gobain
K-LL075 T-LL075 K-LL075
MFLX40619-96EA 3097 MXN
MFLX40619-96 MFLX31200-50 MFLX31201-50
Saint Gobain® Plastic Luer-to-Sanitary Adapters
Racores y conectores
Attach with just a twist!
  • Choose between polypropylene (PP) and PVDF material options
Standard connection type for syringes, plus ideal for many applications with small tubing. These Female luer lock x 3/4" mini sanitary adapters are available in either polypropylene or PVDF.