Pentair In-line Cartridge Filtration Systems
Proveedor: Pentair
255637-43 255576-43 255632-43 255579-43 255582-43
MFLX01507-63EA 412 MXN
MFLX01507-63 MFLX01507-64 MFLX01507-87 MFLX01507-65 MFLX01507-84
Pentair In-line Cartridge Filtration Systems
Filtros Sistemas de filtro de cartucho
A complete filtration solution in one unit.
  • Include a pre- and post- filter
  • A variety of sizes to meet your capacity needs
  • Ideal choice for post-RO, under-sink, ice-maker, and food service applications
Each in-line filter includes an injection-molded polypropylene body and cap, and pre- and post-filters. Choose from either FPT connections or John Guest® push-to-connect fittings. Two unique filter types are available — for the removal of chlorine taste and odor, or removal of rust and sediment. All filters have a maximum temperature rating of 100 °F (38 °C) and 125 psi pressure rating. Filters meet NSF standards.