PharmaLok Series Single-Use series Clamp
CPAX150-U71
MFLX15958-10EA 3560.4 MXN
MFLX15958-10
PharmaLok Series Single-Use series Clamp
Abrazaderas Pinzas para uso general
Single-handed closure- a significant advancement in Biopharma industry.
  • Easy to use – No tools needed
  • - ASME BPE 2014 DT-9.3.1 compliant , DIN compliant
  • - Connects both plastic & stainless steel fittings
  • - SIP/CIP compatible
PharmaLok’s Patent pending design allows effortless attachment of fittings to bag ports and filters. The user simply rests the fitting in the PharmaLok’s regulation-sized slot, lightly grips the surrounding handles, and listens for the sound of clicks to confirm a secure connection.
Specifications:Open clamp sizes include .5” to .75“ and 1” to 1.5“ Tube clamp supports tubing sizes : .5” to 1.25“Operating Temperature: -94 °F (-70 °C) to 140 °F (60 °C)Maximum Pressure: 80 psi (conforms to ASME BPE-2014 PM-4.2.2)