SP Bel-Art Sterileware® Sterile Mini Tongs, Bel-Art Products, a part of SP
Proveedor: Bel-Art
Sterileware®
F37944-0000
47733-014CS 1393.99 MXN
47733-014
SP Bel-Art Sterileware® Sterile Mini Tongs, Bel-Art Products, a part of SP
Pinzas de crisol
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals.
  • Lightweight
  • One-piece polystyrene plastic construction
  • Ridged jaws securely hold wet or dry samples
  • Individually packaged
  • Sterilized via gamma irradiation
Their ridged jaws securely hold wet or dry samples
Tongs are individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization.
One-piece, sterile tongs are made from polystyrene plastic.
Información para pedidos: Individually wrapped and packed for one-time use and disposal.