VWR® Beaker Tongs, Plastic-Coated Tips
Proveedor: VWR
VWR®
11648-100
82027-360CS 33719.07 MXN
82027-360
VWR® Beaker Tongs, Plastic-Coated Tips
Pinzas de crisol
Tongs feature carbon steel, cadmium-plated construction with plastic-coated tips.
  • Carbon steel, cadmium-plated construction
  • Plastic-coated tips
Designed for the safe and simple handling of beakers and similar vessels from 50 to 2000 mL capacities.