VWR® Tongs, PTFE-Coated
Proveedor: VWR
VWR®
11648-108
82027-376CS 26070.66 MXN
82027-376
VWR® Tongs, PTFE-Coated
Pinzas de crisol
Designed to handle hot, cold, corrosive, or sticky materials without scratching or damaging container exteriors.
  • Stainless steel construction
  • PTFE coating
  • Nonsticking and non-corrosive
Contoured jaw enables the handling of crucibles and other objects. Tongs are used for handling chemically active materials.