VWR® Beaker Tongs, Fiberglass Coating
Proveedor: VWR
VWR®
11648-107
82027-374CS 61415.97 MXN
82027-374
VWR® Beaker Tongs, Fiberglass Coating
Pinzas de crisol
Tongs feature a stainless steel construction, a riveted joint, and fiberglass coating for use in high-temperature applications.
  • Stainless steel construction
  • For use in high-temperature applications
Designed for the safe and simple handling of beakers and similar vessels from 50 to 2000 mL capacities.