VWR® Micro Spatulas, Color-Coded Handles
Proveedor: VWR
VWR®
3300
57951-010PK 1054.87 MXN
57951-010
VWR® Micro Spatulas, Color-Coded Handles
Espátulas
Use to dispense/transfer expensive chemicals, place particulate matter in weighing containers, apply micro amounts of adhesive for tacking down specimens, or mix small granular samples.
  • Best tool for all micro samples
  • Four handy sizes in each pack
  • Stainless steel blades
  • Color-coded handles
Micro spatulas are perfect for handling extremely small samples. Spatulas have flat-ended, stainless steel blades. Pack of four includes four different blade widths: 0.1, 0.3, 0.5, and 1.0 mm. (0.004, 0.01, 0.02, and 0.04"). Handles are color-coded in blue, white, green, and red for easy size identification. Appropriate for use with balances, microscopes, or combustion analyzers.