VWR® Rochester-Pean Hemostatic Forceps
Proveedor: VWR
VWR®
11648-130 11648-132 11648-129 11648-131
82027-418CS 8029.56 MXN
82027-418 82027-424 82027-420 82027-422
VWR® Rochester-Pean Hemostatic Forceps
Pinzas
Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
  • Stainless steel construction
  • Reusable
  • Non-sterile
  • Premium grade
Horizontal serrations help ensure forceps remain clamped tight.