SP Bel-Art Test Tube Baskets, Polypropylene, Bel-Art Products, a part of SP
Proveedor: Bel-Art
F18737-0010 F18738-0010 F18739-0010 F18737-0000 F18739-0000 F18738-0000
60914-208CS 4229.24 MXN
60914-300 60914-208 60914-263 60914-285 60914-241 60914-220
SP Bel-Art Test Tube Baskets, Polypropylene, Bel-Art Products, a part of SP
Cestas Cestas, tubos de ensayo
Ideal for rinsing glassware, these polypropylene baskets are available in three sizes with or without lids.
  • Polypropylene basket construction
  • Easy to drain
  • Autoclavable
  • Available with or without lids
Simply invert lidded baskets for easy draining. Steam autoclavable at 121ºC (250ºF).