Nalgene® Autoclaving Baskets, Polypropylene, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
6917-0127 6917-0230 6917-0150
60915-392CS 6168.53 MXN
60915-390 60915-392 60915-394
Nalgene® Autoclaving Baskets, Polypropylene, Thermo Scientific
Cestas Cestas, laboratorio
These baskets hold labware during autoclaving and washing
They allow complete venting and draining. Smooth surface will not scratch plastic or glass. White polypropylene will not rust. Nestable for storage.