VWR® Hayman Style Spatulas, Stainless Steel
Proveedor: VWR
VWR®
11648-182 11648-181
82027-524CS 59094.36 MXN
82027-524 82027-522
VWR® Hayman Style Spatulas, Stainless Steel
Espátulas
Stainless steel spatulas ideal for applications involving the transfer or mixing of small sample amounts.
  • Efficient design
  • Stainless steel construction
Model 82027-522 features a round spatula on one end and a spoon on the other. Model 82027-524 features a tapered, bent-tip spatula on one end and a spoon on the other. Efficient design reduces the risk of sample loss.