Baskets, Aluminum, Black Machine Company
Proveedor: Black Machine Company
A300/A A301/C
60905-331EA 508.06 MXN
60905-331 60905-116
Baskets, Aluminum, Black Machine Company
Cestas Cestas, laboratorio
These lightweight aluminum test tube baskets are made of heavy gauge, expanded aluminum that is riveted for long life and years of rust-free service
Strip bands cover sharp edges for safe handling and a smooth appearance. Expanded metal construction provides rigidity and full flow-through of the cleaning solution.
Información para pedidos: Manufactured in the USA. Custom baskets are available. Contact your Avantor sales representative for more information.