Baskets, Epoxy-Coated Aluminum, Black Machine Company
Proveedor: Black Machine Company
E101/F E101/C E100/C E101/H E101/B
60905-380EA 1153.9 MXN
60905-380 60905-374 60905-372 60905-378 60905-375
Baskets, Epoxy-Coated Aluminum, Black Machine Company
Cestas Cestas, laboratorio
These economical baskets hold laboratory ware for washing, storage, or autoclaving
They hold up under many laboratory environments, withstanding broad temperature ranges (–220 to 120°C), autoclaving, or ice/water conditions. Epoxy-coated aluminum baskets resist rusting, corrosion, abrasion, solvents, acids, and alkalis. Flow-through design permits easy cleaning and rinsing of many pieces of labware at one time.
Información para pedidos: Manufactured in the USA. Custom baskets are available. Contact your Avantor sales representative for more information.