VWR® Lab Scoop, Stainless Steel
Proveedor: VWR
VWR®
11648-170 11648-169
82027-498CS 3049.2 MXN
82027-498 82027-500
VWR® Lab Scoop, Stainless Steel
Palas
The rounded tip of this scoop makes it easy to push into powders or break up solidified chemicals into smaller pieces, while its concave shape makes it ideal for picking them up.
  • Sturdy stainless steel construction
  • Handle and scoop sold separately
Lab scoops are sold in separate handle and scoop components to readily replace parts inexpensively.