Beta Containers, Mitchell Plastics™
Proveedor: Mitchell Plastics
RP-300 RP-400
56615-902EA 5851.2 MXN
56615-900 56615-902
Beta Containers, Mitchell Plastics™
Cajas Cajas de radiación beta y gama
Containers enable safe storage of reagents that emit beta isotopes such as 3H, 14C, 32P, and 35S
  • Safe storage of reagents that emit beta isotopes such as 3H, 14C, 32P, and 35S
  • 9.5mm (3/8") acrylic walls provide protection from exposure to beta radiation during transport, storage, or general use
  • Withstands temperatures as low as –70°C (–94°F)
  • Hinged lids provide easy access to interior
The containers can withstand temperatures as low as –70°C (–94°F). The hinged lids provide easy access to interior.
Their 9.5mm (3/8") thick acrylic walls provide protection from exposure to beta radiation during transport, storage, or general use.