Mottler Brushes, Mill-Rose
Proveedor: Mill-Rose
75114B 75111B 75112B
17230-027EA 168.21 MXN
17230-027 17230-049 17230-060
Mottler Brushes, Mill-Rose
Cepillos
This soft, thick goat hair brush is appropriate for dusting balances and weights
The non-scratching brush head is held by a wide metal ferrule riveted to a hardwood handle.