Nalgene® Carboys with Spigot and Handles, Polypropylene, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
2319-0020 2319-0050 2319-0130
16101-404CS 34559.63 MXN
16101-404 16101-448 16101-481
Nalgene® Carboys with Spigot and Handles, Polypropylene, Thermo Scientific
Frascos Garrafones
Use for storing and dispensing solutions and media.
  • Provide excellent chemical resistance
Quick Action spigot boss is molded in one piece directly to the body of the carboy for strength and leak-resistance. Cap size: 83B. Also takes No. 13¹/₂ stopper.
For replacement caps, see cap 16225-234 in 16219-122 series.
Note: Autoclave using a properly vented closure. Disassemble spigot before autoclaving.
Ideal for sterile water storage, carboys can be autoclaved before refilling to stop recurring algae or bacterial growth. Graduated to deliver in 1 gal. and 5 L increments.
Información para pedidos: Supplied with carboy, polypropylene spigot, and screw cap.