VWR® Tweezers, PTFE
Proveedor: VWR
VWR®
027.101 027.150 027.151 027.100
89026-426EA 1794.96 MXN
89026-418 89026-426 89026-424 89026-420
VWR® Tweezers, PTFE
Pinzas
Tweezers have a smooth PTFE non-stick surface.
  • PTFE construction
  • Non-contaminating
  • Inert
For use in temperatures of 0 to 280°C (32 to 540°F). Non-contaminating and inert to virtually all chemical reagents.