Cylinder and Bottle Brushes, Radial End, Mill-Rose
Proveedor: Mill-Rose
3065B 72065B 72063B 72064B
70230-018EA 63.02 MXN
70230-018 17066-089 17066-058 17066-080
Cylinder and Bottle Brushes, Radial End, Mill-Rose
Cepillos
For washing cylinders, large test tubes and bottles.
  • Twisted wire handle with hanging loop
  • Full radial end provides thorough cleaning action
Available in black natural bristles or black nylon filaments.