VWR® Microtube Storage Boxes
Proveedor: Avantor
VWR®
R8300-P-VWR R8300-Y-VWR R8300-B-VWR R8300-G-VWR R8300-VWR R8300-A-VWR R8300-BLK-VWR R8300-O-VWR
40000-324CS 7885.23 MXN
40000-324 40000-326 40000-320 40000-332 40000-322 40000-330 40000-328 89084-934
VWR® Microtube Storage Boxes
Cajas Criocajas
These durable, polypropylene boxes maintain their integrity under harsh freezer conditions and will not absorb water or degrade like cardboard storage boxes.
  • Racks for the chest freezers store up to nine boxes
  • Racks for the upright freezers store up to eight boxes
  • Each box holds one hundred 1.5 mL or 2.0 mL tubes
  • Aailable in seven colors (with clear lids)
  • Autoclavable to 121 °C (249.8 °F)
  • Temperature Range: –80 to 121 °C (–112 to 249.8 °F)
A three-point hinge and positive catch provide quick access to samples and keep them secure when box is closed. Alphanumeric indexing is printed inside the boxes which aids in sample identification.
Información para pedidos: Assorted boxes includes one box in each of the following colors: blue, green, orange, pink, and yellow.