VWR® Freezer Boxes
Proveedor: Avantor
VWR®
2709-229-000 2709-220-000 2708-229-000
16466-020PK 821.83 MXN
16466-020 16466-022 16466-024
VWR® Freezer Boxes
Cajas Criocajas
VWR® Freezer Boxes are made of polypropylene and come with an alphanumerically labeled grid for identifying contents.
  • Boxes are autoclavable
  • Freezable to -90°C (-130°F)
The 50- and 100-place freezer boxes come with attached hinged lid and snap lock closure for versatility. They are designed to hold 1.5 mL and 2.0 mL tubes and some 1.2 mL cryo vials. Indented lids can be labeled. The 81-place box is designed to fit into existing freezer storage racks, holds 1.5 or 2.0 mL microcentrifuge tubes, and includes an attached clear lid.
Información para pedidos: Assorted colors packages include one each in blue, green, orange, red, and yellow.