Volumetric Flask Brushes, Mill-Rose
Proveedor: Mill-Rose
72131W 72130W 72132W
70230-014EA 84.87 MXN
70230-014 70230-010 70230-012
Volumetric Flask Brushes, Mill-Rose
Cepillos
Made of white natural bristles mounted on a durable, plastic handle.
Flexible, curved brush adapts to the shape of flasks.