Buret and Pipet Brushes, Mill-Rose
Proveedor: Mill-Rose
72212W 72210W 72211W
17110-069PK 280.02 MXN
17110-069 17110-047 17110-025
Buret and Pipet Brushes, Mill-Rose
Cepillos
Extra long flexible wire handles reach into burets, pipets, and long tubing for cleaning.
Unbleached white bristles on galvanized wire with fan-style tip to protect against scratching.