VWR® Freezer Storage Boxes for 50 and 15 ml Centrifuge Tubes, 16 and 36-Place
Proveedor: Avantor
VWR®
2703-220-000 2704-220-000
83009-692PK 821.83 MXN
83009-692 82017-232
VWR® Freezer Storage Boxes for 50 and 15 ml Centrifuge Tubes, 16 and 36-Place
Cajas Criocajas
Polypropylene storage boxes are designed to hold 50 mL or 15 mL VWR® SuperClear tubes for long-term storage of frozen samples.
  • Boxes maintain integrity under harsh freezer conditions; down to -90°C (-130°F)
  • Clear cover is included
  • Autoclavable to 122°C (251.6°F)
Boxes feature a white upper body, which can be easily labelled with a lab marker, and a numerical reference grid.