SP Bel-Art Autoclavable Carboys, Polypropylene, Bel-Art Products, a part of SP
Proveedor: Bel-Art
SCIENCEWARE®
F10794-0025 F11846-0025 F11846-0050 F10794-0050
47751-718EA 4499.6 MXN
47751-718 47751-716 74950-052 47751-722
SP Bel-Art Autoclavable Carboys, Polypropylene, Bel-Art Products, a part of SP
Frascos Garrafones
Rugged carboys for direct contact with foods and beverages, and are USP Class VI certified for the demands of the biotechnology and pharmaceutical industries.
  • Excellent chemical resistance
  • Large handles
  • Polypropylene construction
  • Autoclavable
  • Easy-to-use spigot
Ideal for storing and dispensing large batches of culture media, distilled water and other solutions, they have excellent chemical resistance and large handles that help with transport.
All components are FDA-grade polypropylene construction and are steam autoclavable at 121 °C (250 °F). When autoclaving, do not engage cap threads. The 83 mm screw cap includes a built-in thermo-plastic elastomer washer for positive seal. Molded-in bung accepts the easy-to-use spigot and is located low on the carboy for more complete drainage.
Certificaciones: 21 CFR 177.1520 compliant.
Información para pedidos: Available with or without spigot.