Nalgene® High-Density Polyethylene Jerricans, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
2240-0015 2240-0050 2243-0013 2243-9013 2240-0025
36382-022CS 13556.74 MXN
36382-022 36382-066 73520-057 36382-044 73520-059
Nalgene® High-Density Polyethylene Jerricans, Thermo Scientific
Frascos Bidones
Sturdy, leakproof containers are intended for hard use.
A wide stance and low center of gravity provide greater stability. Integral spout is long enough for accurate pouring, and features a strap-fastened cap. Molded-in graduations are provided in liters and gallons. Recessed bottom provides second handgrip for pouring. Jerricans 73520-057 and -059 weigh 13 kg (28.7 lbs.) at their normal fill volume, just right for unassisted lifting. Jerrican 73520-059 features a second spout with a tethered cap for vented pouring. This provides access for refilling when the 53B spout is fitted with connections to equipment.
Información para pedidos: Jerricans are supplied with 53B (buttress-style) polypropylene closures. Jerrican 73520-059 also comes with a 38-430 polypropylene closure.