Nalgene® Graduated Griffin Beakers, PMP, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
1203-0050 1203-1000 1203-4000 1203-0600 1203-2000 1203-0150 1203-0400 1203-0100 1203-0250
13918-043CS 5589.18 MXN
13918-043 13918-087 13918-065 13918-167 13918-145 13918-189 13918-123 13918-101 13918-203
Nalgene® Graduated Griffin Beakers, PMP, Thermo Scientific
Vasos de precipitados
Shatter-resistant PMP (polymethylpentene) has glass-like transparency, top heat tolerance and chemical resistance.
  • Non-toxic, non-contaminating, biologically inert
  • Safe for repeated autoclaving
  • Easy-to-read, silk-screened graduations, plus maximum temperature use, “no flame” symbol and resin code
  • Oval marking area, tapered walls
Takes constant usage at 150°C (302°F); intermittent at 175°C (347°F).
Certificaciones: Meet ISO/DIS 7056 international standards for laboratory plasticware accuracy.