Nalgene® Polysulfone Dilution Bottles, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
2500-0380 2500-0280
16161-847CS 18508.64 MXN
16161-847 16161-803
Nalgene® Polysulfone Dilution Bottles, Thermo Scientific
Frascos Botellas para dilución
Transparent polysulfone bottle with linerless polypropylene caps. Appropriate for most aqueous and many organic compounds.
  • French square design with sloping shoulders
  • Closures withstand from 0 to 135 °C (32 to 275 °F)
  • Bottles withstand temperatures from –100 to 165°C (–148 to 329 °F)
  • Guaranteed leakproof
Graduations at both 90 and 99 ml, guaranteed accurate to ±1 ml, for easy preparation of ¹/₁₀ or ¹/₁₀₀ dilutions in same bottle. Both bottle and cap material is nontoxic and safe for food; autoclavable but use with Tween® will reduce bottle life.
For replacement caps, see 16219-030 series.
Información para pedidos: Supplied with linerless polypropylene screw closure. Bottles available in narrow mouth and wide mouth styles.
Precaución: Before autoclaving, set cap on top of bottle without engaging threads.