VWR®, Griffin Low Form Beakers with Double-Capacity Scale, Borosilicate Glass
Proveedor: Avantor
VWR®
VW14000-20 VW14000-250 VW14000-4000
89000-204CS 3445.86 MXN
89000-204 89000-218 89000-194
VWR®, Griffin Low Form Beakers with Double-Capacity Scale, Borosilicate Glass
Vasos de precipitados
Beakers feature a thick, slightly flared, beaded top with a spout designed to have excellent pouring characteristics.
  • Thick, slightly flared, beaded top with a spout designed to have excellent pouring characteristics
  • Uniform walls and are made of Type I, Class A borosilicate glass
  • Feature a durable matte finish marking area suitable for use with an ordinary pencil
They have uniform walls and are made of Type I, Class A borosilicate glass. Beakers 20 mL and larger are printed with an easy-to-read white graduated scale for approximate measurement. All beakers feature a durable matte finish marking area suitable for use with an ordinary pencil.