KIMAX® Heavy-Duty Beakers, Low Form, Double Scale, Borosilicate Glass, Kimble Chase
Proveedor: DWK Life Sciences
KIMAX®
14005-1000 14005-600 14005-250 14005-4000 14005-2000 14005-400
89001-076CS 9949.8 MXN
13913-480 89001-078 89001-076 89001-074 89001-084 89001-081 89001-082
KIMAX® Heavy-Duty Beakers, Low Form, Double Scale, Borosilicate Glass, Kimble Chase
Vasos de precipitados
Beakers feature thick, uniform walls throughout and an extra-thick, evenly tooled top rim
Spout is designed for optimum performance. Heavy construction is intended to prolong life expectancy with increased and more rigorous usage. All sizes have a durable matte finish marking area and a white graduated scale.
Certificaciones: Designed from ASTM Specification E960, Type II requirements.