Kartell® Graduated Beakers, Low Form, PMP
Proveedor: Kartell
VWR®
K1723-VWR K1728-VWR K1721-VWR K1720-VWR K1724-VWR K1726-VWR K1725-VWR K1722-VWR
89044-520CS 4911.24 MXN
89044-520 89044-510 89044-522 89044-516 89044-518 89044-508 89044-512 89044-514
Kartell® Graduated Beakers, Low Form, PMP
Vasos de precipitados
Crystal-clear beakers feature excellent chemical resistance and a large spout for efficient pouring.
  • Autoclavable
  • Molded from polymethylpentene (PMP)
  • Can withstand a temperature of up to 170°C (338°F) for short periods
Permanent blue graduations are divided into finer increments for more precise measuring.
Certificaciones: Beakers conform to ISO 7056 1981 (E) and BS 5404 Part 1.