VWR® Freezer Storage Boxes
Proveedor: VWR
VWR®
2710-220-300 2708-220-000 2711-220-300 2712-220-300
89004-562CS 3348.14 MXN
89004-562 89004-564 16466-018 89004-566
VWR® Freezer Storage Boxes
Cajas Criocajas
Transparent polycarbonate freezer boxes can hold 0.5, 1.5, 2, and 5 mL freestanding screw cap tubes.
  • Polycarbonate fabrication
  • Numbered grid
They feature a numbered grid for easy tube location and a transparent lid that does not require orientation to close. Boxes come in three sizes and fit most freezer box racks. Autoclavable.
Min. Temperature: –190 °C (–310 °F)