VWR® Freezer Box for Microcentrifuge Tubes
2713-220-300
89004-568CS 9850.9 MXN
89004-568
VWR® Freezer Box for Microcentrifuge Tubes
Polycarbonate freezer box holds one hundred 0.5, 1.5, or 2.0 mL freestanding screw cap microcentrifuge tubes and features numbered posts for easy location of samples.
  • Polycarbonate construction
  • Suitable over a range of temperatures
Drain holes are provided in box for use in temperatures as low as -190°C (-310°F). Transparent lid provides a clear view of all samples, and does not require orientation to close. Autoclavable. Translucent purple with clear lid.