


Antimony (III) chloride ≥99% BAKER ANALYZED® ACS
JT0878-1
: Avantor
:
- Pack type:Glass bottle, black T/E cap
- Size:500 g
- Cat. no.:JT0878-1
- Supplier No.:0878-01
- Assay (SbCl₃) (by iodometry):≥99.0 %
- Insoluble in Chloroform:≤0.05 %
- Sulfate (SO₄):≤0.005 %
- Arsenic (As):≤0.02 %
- Calcium (Ca):≤0.005 %
- Copper (Cu):≤0.001 %
- Iron (Fe):≤0.002 %
- Lead (Pb):≤0.005 %
- Potassium (K):≤0.01 %
- Sodium (Na):≤0.02 %
:
:228.12 g/mol
:13,00713
:223.5 °C (1013 hPa)
:223.5 °C (1013 hPa)
:73 °C
:3.14 g/cm³ (20 °C)
:Ambient
:233-047-2
:1733
:8,II
:Management of Change (MOC) category = N
:GHS05
:GHS07
:GHS09
:10025-91-9
:SbCl₃
:MFCD00011212