Dropping Bottles, Amber, Kimble®, DWK Life Sciences
Proveedor: DWK Life Sciences
15040P-60 15040G-60 15040P-30 15040G-30
16199-039CS 677.84 MXN
16199-039 16199-037 16199-034 16199-031
Dropping Bottles, Amber, Kimble®, DWK Life Sciences
Frascos Frascos de goteo
ASTM E438, Type III amber glass protects light-sensitive chemicals.
Supplied with 20-400 GPI screw cap, bulb and dropper assembled on the bottle. Dropper is available in polyethylene or USP Type III glass.