Glass Dropping Bottles, Wheaton®, DWK Life Sciences
Proveedor: DWK Life Sciences
W211755 W211735 W211754 W211734
14216-234CS 3896.37 MXN
14216-232 14216-234 14216-236 14216-230
Glass Dropping Bottles, Wheaton®, DWK Life Sciences
Frascos Frascos de goteo
With glass hood stopper.
Channel in stopper allows drop-by-drop delivery of contents when bottle is tipped. A partial turn of the stopper seals bottle. Available in clear and amber glass. USD Type III Glass.