KIMAX® Culture Flasks, Capacity Scale, Kimble Chase
Proveedor: DWK Life Sciences
KIMAX®
25615-1000 25615-2000 25615-125 25615-250 25615-500
89001-296CS 7426.3 MXN
89001-300 89001-298 89001-296 89000-780 89001-302
KIMAX® Culture Flasks, Capacity Scale, Kimble Chase
Matraces Matraces de cultivo de células
Flasks have a thick-walled, long tubing neck with a plain fire-polished finish sealed to an Erlenmeyer-shaped body
Designed from ASTM Specification E1404, Type IV, Class 1 requirements.
Neck is long enough to accept a metal or plastic cap or a cotton plug. Durable white ceramic enamel scale indicates approximate volume.