KIMAX® Baffled Culture Flasks, Kimble Chase
Proveedor: DWK Life Sciences
KIMAX®
25630-125 25630-500 25630-2000 25630-250 25630-1000
89000-986CS 3186.92 MXN
89000-986 89000-988 89000-990 89000-992 89000-984
KIMAX® Baffled Culture Flasks, Kimble Chase
Matraces Matraces de cultivo de células
Designed for use with rotary and reciprocating shakers where baffle indents provide greater turbulence to improve oxygen transfer rates or aeration
Flask has a neck with a plain fire-polished finish suitable for a metal cap, plastic cap, foam plug or cotton plug.
Información para pedidos: Closures are available separately.