Rubbermaid® Commercial Food/Tote Box Lids, Essendant
Proveedor: Essendant
Rubbermaid®
RCP3302CLE RCP3302CLE RCP3310CLE RCP3502WHI RCP3510WHI
62660-429EA 445.57 MXN
62660-620 62660-825 62660-429 62660-824 62660-949
Rubbermaid® Commercial Food/Tote Box Lids, Essendant
Artículos para la manipulación de alimentos
This bulk food storage container lid reduces risk of cross-contamination, secures storage while stacked, and can be used as tray when displayed upside down.
  • Crystal-clear material provides visibility, durability and stain-resistance
  • Tight-fitting, snap-on lid with stacking ribs keeps food secure
  • Date control panels for easy inventory control
The Rubbermaid Commercial food storage lid for food tote box helps reduce food spoilage costs.
Certificaciones: FDA Compliant.