KIMBLE® KIMAX® Gas Washing Bottles, Tall Form, [ST] Stopper, DWK Life Sciences
Proveedor: DWK Life Sciences
KIMAX®
15060-125 15060-250 15060-500
89001-572CS 3530.2 MXN
89001-572 89001-570 89001-568
KIMBLE® KIMAX® Gas Washing Bottles, Tall Form, [ST] Stopper, DWK Life Sciences
Frascos Botellas de lavado de gases
For washing or drying gases.
Gas flows into the bottom of the bottle through the center tube, rises through the washing or drying medium, and exits through the side tube of the stopper. Bottles have [ST] 34/28 stoppers and inlet and outlet ports that accept 0.63 cm (¹/₄") Int.Ø flexible tubing.